Learn More
ACROBiosystems Inc Recombinant Protein;Human CD300LG / Nepmucin Protein;HEK293;100UG;CDG,CD300LG,CLM9,TREM4,CD300g,Nepmucin,CLM-9,TREM-4
Numéro de catalogue. 501658942
Encompass
Recombinant Protein;Human CD300LG / Nepmucin Protein;100UG;Human CD300LG, His Tag (CDG-H5223) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 247 (Accession # AAH25395).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.6 kDa. The protein migrates as 33-68 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.
Numéro de catalogue. 501658942 Fournisseur ACROBiosystems Inc Code fournisseur CDGH5223100UG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant Protein;Human CD300LG / Nepmucin Protein;100UG;Human CD300LG, His Tag (CDG-H5223) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 247 (Accession # AAH25395).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.6 kDa. The protein migrates as 33-68 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.