Recombinant Protein;100UG;Human CD28H, His Tag (CDH-H52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Gly 150 (Accession # Q96BF3-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.9 kDa. The protein migrates as 22-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD28H, His Tag (Cat. No. CDH-H52H3) at 5 g/mL (100 L/well) can bind Biotinylated Human B7-H7, Fc Tag, Avi Tag (Cat. No. B77-H82F5) with a linear range of 10-156 ng/mL (QC tested).;Immobilized Human CD28H, His Tag (Cat. No. CDH-H52H3) at 5 g/mL (100 L/well) can bind Human B7-H7, Fc Tag (Cat. No. B77-H5257) with a linear range of 1-78 ng/mL (Routinely tested).