Learn More
ACROBiosystems Inc ACROBiosystems Human CD209 / DC-SIGN Protein, His Tag
Catalog No. 501956467
Encompass
Recombinant Protein;100UG;Human CD209, His Tag (CD9-H5246) is expressed from human 293 cells (HEK293). It contains AA Gln 59 - Ala 404 (Accession # Q9NNX6-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 41.3 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 501956467 Supplier ACROBiosystems Inc Supplier No. CD9H5246100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;100UG;Human CD209, His Tag (CD9-H5246) is expressed from human 293 cells (HEK293). It contains AA Gln 59 - Ala 404 (Accession # Q9NNX6-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 41.3 kDa. The protein migrates as 40-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.