Learn More
ACROBiosystems Inc Recombinant Protein;Biotinylated Human CD200 R1 / CRTR2, Fc Tag, Avi Tag (Avitag™);HEK293;25UG;CR2,CD200 R1,CD200R,CRTR2,MOX2R,OX2R
Catalog No. 501658235
Encompass
Recombinant Protein;Biotinylated Human CD200 R1 / CRTR2, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human CD200 R1, Fc Tag (CR2-H82F4) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Leu 266 (Accession # AAQ19772.1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 55.8 kDa. The protein migrates as 70-115 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD200, Fc Tag (Cat. No. OX2-H5251) at 2 g/mL (100 L/well) can bind Biotinylated Human CD200 R1, Fc Tag (Cat. No. CR2-H82F4) with a linear range of 0.5-7.8 ng/mL (QC tested).
Catalog No. 501658235 Supplier ACROBiosystems Inc Supplier No. CR2H82F425UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;Biotinylated Human CD200 R1 / CRTR2, Fc Tag, Avi Tag (Avitag™);25UG;Biotinylated Human CD200 R1, Fc Tag (CR2-H82F4) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Leu 266 (Accession # AAQ19772.1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 55.8 kDa. The protein migrates as 70-115 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD200, Fc Tag (Cat. No. OX2-H5251) at 2 g/mL (100 L/well) can bind Biotinylated Human CD200 R1, Fc Tag (Cat. No. CR2-H82F4) with a linear range of 0.5-7.8 ng/mL (QC tested).