Recombinant Protein;1MG;Human CD2, Fc Tag (CD2-H5258) is expressed from human 293 cells (HEK293). It contains AA Lys 25 - Asp 209 (Accession # AAH33583).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 47.7 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD58, His Tag (Cat. No. LF3-H5225) at 10 g/mL (100 L/well) can bind Human CD2, Fc Tag (Cat. No. CD2-H5258) with a linear range of 5-156 ng/mL (QC tested).