Learn More
ACROBiosystems Inc Recombinant Protein;Mouse CD155 / PVR Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;50UG;CD5,CD155,PVR,FLJ25946,PVS,TAGE4,HVED,NECL5,NECL-5,TAGE-4
Catalog No. 501658288
Encompass
Recombinant Protein;Mouse CD155 / PVR Protein, Mouse IgG2a Fc Tag, low endotoxin;50UG;Mouse CD155, mouse IgG2a Fc tag (CD5-M5254) is expressed from human 293 cells (HEK293). It contains AA Asp 29 - Leu 348 (Accession # Q8K094-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 61.8 kDa. The protein migrates as 35 kDa and 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse TIGIT, His Tag (Cat. No. TIT-M52E6) at 10 g/mL (100 L/well) can bind Mouse CD155, mouse IgG2a Fc tag (Cat. No. CD5-M5254) with a linear range of 0.039-1.25 g/mL (QC tested).
Catalog No. 501658288 Supplier ACROBiosystems Inc Supplier No. CD5M525450UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;Mouse CD155 / PVR Protein, Mouse IgG2a Fc Tag, low endotoxin;50UG;Mouse CD155, mouse IgG2a Fc tag (CD5-M5254) is expressed from human 293 cells (HEK293). It contains AA Asp 29 - Leu 348 (Accession # Q8K094-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 61.8 kDa. The protein migrates as 35 kDa and 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse TIGIT, His Tag (Cat. No. TIT-M52E6) at 10 g/mL (100 L/well) can bind Mouse CD155, mouse IgG2a Fc tag (Cat. No. CD5-M5254) with a linear range of 0.039-1.25 g/mL (QC tested).