Learn More
ACROBiosystems Inc Recombinant Protein;Rhesus macaque CD155 / PVR Protein;HEK293;1MG;CD5,CD155,PVR,FLJ25946,PVS,TAGE4,HVED,NECL5,NECL-5,TAGE-4
Catalog No. 501657600
Encompass
Recombinant Protein;Rhesus macaque CD155 / PVR Protein;1MG;Rhesus macaque CD155 (CD5-R52H5) is expressed from human 293 cells (HEK293). It contains AA Asp 28 - Asn 343 (Accession # Q0MSE6-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.3 kDa. The protein migrates as 48-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rhesus macaque CD155 (Cat. No. CD5-R52H5) at 5 g/mL (100 L/well) can bind Cynomolgus / Rhesus macaque TIGIT, Fc Tag (Cat. No. TIT-C5254) with a linear range of 0.02-0.313 g/mL (QC tested).;Immobilized Rhesus macaque CD155 (Cat. No. CD5-R52H5) at 5 g/mL (100 L/well) can bind Biotinylated Human DNAM-1, Fc Tag (Cat. No. DN1-H82F9) with a linear range of 10-156 ng/mL (Routinely tested).
Catalog No. 501657600 Supplier ACROBiosystems Inc Supplier No. CD5R52H51MG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;Rhesus macaque CD155 / PVR Protein;1MG;Rhesus macaque CD155 (CD5-R52H5) is expressed from human 293 cells (HEK293). It contains AA Asp 28 - Asn 343 (Accession # Q0MSE6-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.3 kDa. The protein migrates as 48-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Rhesus macaque CD155 (Cat. No. CD5-R52H5) at 5 g/mL (100 L/well) can bind Cynomolgus / Rhesus macaque TIGIT, Fc Tag (Cat. No. TIT-C5254) with a linear range of 0.02-0.313 g/mL (QC tested).;Immobilized Rhesus macaque CD155 (Cat. No. CD5-R52H5) at 5 g/mL (100 L/well) can bind Biotinylated Human DNAM-1, Fc Tag (Cat. No. DN1-H82F9) with a linear range of 10-156 ng/mL (Routinely tested).