Learn More
ACROBiosystems Inc ACROBiosystems Human CD155 / PVR Protein, His Tag
Catalog No. 501957504
Encompass
Recombinant Protein;100UG;Human CD155, His Tag (CD5-H5223) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 343 (Accession # NP_006496.4).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.9 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 501957504 Supplier ACROBiosystems Inc Supplier No. CD5H5223100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;100UG;Human CD155, His Tag (CD5-H5223) is expressed from human 293 cells (HEK293). It contains AA Trp 21 - Asn 343 (Accession # NP_006496.4).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.9 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.