Learn More
ACROBiosystems Inc Recombinant Protein;Human CD117 / c-kit Protein, Fc Tag;HEK293;100UG;CD7,CD117,SCFR,c-Kit,KIT
Catalog No. 501658967
Encompass
Recombinant Protein;Human CD117 / c-kit Protein, Fc Tag;100UG;Human CD117, Fc Tag (CD7-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Thr 516 (Accession # P10721-2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 81.9 kDa. The protein migrates as 100-125 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD117, Fc Tag (Cat. No. CD7-H5255) at 2 g/mL (100 l/well) can bind Mouse SCF, His Tag (Cat. No. SCF-M5228) with a linear range of 0.05-1.2 ng/mL (Routinely tested).;Immobilized Human CD117, Fc Tag (Cat. No. CD7-H5255) at 2 g/mL (100 l/well) can bind Biotinylated Mouse SCF (Cat. No. SCF-M8228) with a linear range of 0.25-20 ng/mL (QC tested).
Catalog No. 501658967 Supplier ACROBiosystems Inc Supplier No. CD7H5255100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;Human CD117 / c-kit Protein, Fc Tag;100UG;Human CD117, Fc Tag (CD7-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 26 - Thr 516 (Accession # P10721-2).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 81.9 kDa. The protein migrates as 100-125 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD117, Fc Tag (Cat. No. CD7-H5255) at 2 g/mL (100 l/well) can bind Mouse SCF, His Tag (Cat. No. SCF-M5228) with a linear range of 0.05-1.2 ng/mL (Routinely tested).;Immobilized Human CD117, Fc Tag (Cat. No. CD7-H5255) at 2 g/mL (100 l/well) can bind Biotinylated Mouse SCF (Cat. No. SCF-M8228) with a linear range of 0.25-20 ng/mL (QC tested).