missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CD BHK-21 Production Medium

Catalog No. A1627701
Change view
Click to view available options
Quantity:
1 x 10 L
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1627701 1 x 10 L
A1627703 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1627701 Supplier Gibco™ Supplier No. A1627701

CD BHK-21 Production Medium

Gibco™ CD BHK-21 Production Medium is a chemically defined, protein-free cell culture medium. It is designed to support the growth of BHK-21 cells in serum-free suspension culture. This medium is suitable for large-scale manufacturing applications.

CD BHK-21 Production Medium is manufactured as follows:

With

  • L-glutamine
  • Dry powder format

Without

  • Phenol Red
  • Protein hydrolysates

Gibco™ CD BHK-21 Production Medium is the first cell culture medium supporting complete serum-free and protein-free processes for BHK-21 cells. The dry powder format is easy to reconstitute, since it has no hydrolysates in the formulation. Adapting cells from serum-containing media is a rapid and simple procedure. BHK-21 cells can achieve high cell densities and maintain high cell viability in this medium.

Gibco™ CD BHK-21 Production Medium is suitable for large-scale suspension culture. Because it does not require supplementation with serum or other undefined components, Gibco™ CD BHK-21 Production Medium eliminates the risks associated with serum, while allowing you to achieve consistent virus titers and simplifying downstream purification.

cGMP Manufacturing
Gibco™ CD BHK-21 Production Medium is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

Specifications

Cell Line BHK-21
Form Powder
Product Type Production Medium
With Additives Glutamine
Without Additives No Phenol Red
For Use With (Application) Viral Vaccine Production
Quantity 1 x 10 L
Content And Storage Store in refrigerator (2–8°C).
Culture Type Suspension Cell Culture

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.