missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CD 293 AGT™ Medium

Catalog No. 12529012
Change view
Click to view available options
Quantity:
10 kg
10 L
100 L
Format:
Ropak Bucket
Securitainer Capped Vial
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
12529012 10 L Securitainer Capped Vial
12529001 100 L Ropak Bucket
12529003 10 kg Ropak Bucket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12529012 Supplier Gibco™ Supplier No. 12529012
Only null left

CD 293 AGT™ is a protein-free, chemically-defined medium in an easy-to-use granular format.

CD 293 AGT™ is a protein-free, chemically-defined medium in an easy-to-use granular format. CD 293 AGT™ is optimized for the growth of suspension cultures of 293 cells.

AGT = Advanced Granulation Technology

Specifications

Cell Line 293 (HEK)
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Form Powder (AGT)
Product Type CD 293 Medium
Serum Level Serum-free
With Additives Phenol Red, Sodium Pyruvate, Sodium Bicarbonate
Without Additives No Glutamine
Quantity 10 L
Shipping Condition Wet Ice
Endotoxin Level Low (≤10 EU/mL)
Format Securitainer Capped Vial
Included Antibiotics No antibiotics
Reconstituted Volume 10 L
Content And Storage Store in refrigerator (2°C to 8°C) and protect from light.
Culture Type Suspension Cell Culture
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.