missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CD 293 AGT™ Medium

Catalog No. 12529003
Change view
Click to view available options
Quantity:
10 kg
10 L
100 L
Format:
Ropak Bucket
Securitainer Capped Vial
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
12529003 10 kg Ropak Bucket
12529001 100 L Ropak Bucket
12529012 10 L Securitainer Capped Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12529003 Supplier Gibco™ Supplier No. 12529003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CD 293 AGT™ is a protein-free, chemically-defined medium in an easy-to-use granular format.

CD 293 AGT™ is a protein-free, chemically-defined medium in an easy-to-use granular format. CD 293 AGT™ is optimized for the growth of suspension cultures of 293 cells.

AGT = Advanced Granulation Technology

Specifications

Cell Line 293 (HEK)
Classification Animal Origin-free
Form Powder (AGT)
Product Type CD 293 Medium
Serum Level Serum-free
Sterility Non-sterile
With Additives Low Glucose, Sodium Pyruvate, Sodium Bicarbonate, HEPES
Without Additives No Glutamine
Glucose Concentration 4500 mg/L
Quantity 10 kg
Shipping Condition Dry Ice
Endotoxin Level Low
Format Ropak Bucket
Included Antibiotics No antibiotics
Inorganic Salts Calcium, Magnesium, Sodium Phosphate
Purity or Quality Grade Certified
Content And Storage Store at 2°C to 8°C in the dark.
Culture Environment CO2
Culture Type Suspension Cell Culture
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.