missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ CCR5 Antibody Blocking Peptide
Specifications
Specifications
| Gene ID (Entrez) | 1234 |
| Species | Human |
| Product Line | Novus |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-56337 |
| Gene Alias | C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5 FLJ78003, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5 |
| Gene Symbol | CCR5 |
| Quantity | 0.1 mg |
| Research Category | Alzheimers Research, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction |
| Show More |