missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ CCDC160 Recombinant Protein Antigen

Catalog No. NBP23104PEP
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP23104PEP 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP23104PEP Supplier R&D Systems™ Supplier No. NBP231104PEP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human CCDC160. The CCDC160 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 347475
Species Human
Product Line Novus
Purity >80% by SDS-PAGE and Coomassie blue staining
Common Name coiled-coil domain-containing protein 160, coiled-coil domain containing 160
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Formulation PBS and 1M Urea, pH 7.4.
For Use With (Application) This Recombinant Antigen is Only Intended to be Used as a Blocking Agent to Confirm Antibody Specificity with The Corresponding Antibody, Catalog Number NBP2-53851. It is Purified by IMAC Chromatography, and The Expected Concentration is Greater Than 0.5 mg/ml
Gene Symbol CCDC160
Molecular Weight (g/mol) 29 kDa
Preservative No Preservative
Quantity 100 μL
Source E. coli
Show More Show Less