missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ CC2D2B Recombinant Protein Antigen

Catalog No. NP230772PEP
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP230772PEP 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP230772PEP Supplier R&D Systems™ Supplier No. NBP230772PEP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human C10orf131

The CC2D2B Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 387707
Species Human
Product Line Novus
Purity >80% by SDS-PAGE and Coomassie blue staining
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
For Use With (Application) Antibody Competition
Gene Symbol CC2D2B
Label Type Unconjugated
Molecular Weight (g/mol) 27 kDa
Quantity 100 μL
Regulatory Status RUO
Source E. coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-54996. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Storage Buffer PBS and 1M Urea, pH 7.4.
Show More Show Less