missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CBQCA Plus Protein Quantitation Kit
GREENER_CHOICE

Catalog No. a66522
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A66522 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A66522 Supplier Invitrogen™ Supplier No. A66522
Only null left

The CBQCA Plus Protein Quantitation Kit is a specialized tool designed for the accurate quantitation of proteins in solution.

The CBQCA Plus Protein Quantitation Kit is a specialized tool designed for the accurate quantitation of proteins in solution. This kit is a refined version of the CBQCA Protein Quantitation Kit (Cat. No. C6667), with a significant modification: potassium cyanide, a toxic compound, has been replaced with mandelonitrile, a less-toxic alternative. This change does not compromise the kit's performance, maintaining the same level of sensitivity and accuracy as its predecessor.

The CBQCA Plus Protein Quantitation Kit exhibits high sensitivity and is capable of detecting as little as 50 ng of bovine serum albumin (BSA) in an assay volume of 100–200 μL. This sensitivity allows for precise measurements even in low-protein samples. Moreover, the kit is compatible with detergents, a feature that enhances its versatility. This compatibility makes it suitable for accurate quantitation of proteins in the presence of lipids, membrane fractions, or detergents, as well as for lipoproteins and small peptides.

The kit includes all necessary components for performing 300–800 assays, depending on the concentration of substrate used. The resulting fluorescent product is maximally excited at 465 nm and has an emission maxima at 550 nm. It is designed for use with a fluorescence microplate reader, but can also be adapted for use with a standard fluorometer or minifluorometer after dilution of the reaction mixture.

Specifications

Assay Range 50 ng to 150 μg
Content And Storage Sufficient for: 300 to 800 assays

• Component A: ATTO-TAG CBQCA derivatization reagent, 5 mg
• Component B: Dimethylsulfoxide (DMSO), 1 mL
• Component C: Mandelonitrile, 20 mM in 4 mL DMSO
• Component D: Bovine serum albumin (BSA) standard, 4 mg
• BupH Borate Buffer Pack

Store in freezer (-5°C to -30°C) and protect from light.
Assay Fluorescent Protein Assay
Detection Method Fluorescence
Product Type Protein Quantitation Assay
For Use With (Application) Quantitating proteins in solution
For Use With (Equipment) Microplate Reader, Fluorometer
Quantity 1 Kit
Shipping Condition Wet Ice
Format Kit
Excitation/Emission 465/550 nm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.