Learn More
ACROBiosystems Inc ACROBiosystems Human CBLB / RNF56 Protein, His Tag
Catalog No. 501956761
Encompass
Recombinant Protein;100UG;Human CBLB, His Tag (CBB-H5148) is expressed from E.coli cells. It contains AA Ala 39 - Phe 426 (Accession # Q13191-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 47.0 kDa. The protein migrates as 46 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 501956761 Supplier ACROBiosystems Inc Supplier No. CBBH5148100UG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Protein;100UG;Human CBLB, His Tag (CBB-H5148) is expressed from E.coli cells. It contains AA Ala 39 - Phe 426 (Accession # Q13191-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 47.0 kDa. The protein migrates as 46 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.