Learn More
Foxx Life Sciences UN/DOT Tight Head Containers for Shipping, Transport, Storage, and Waste; 20L (5 Gallon), HDPE, 70mm Cap, UN 3H1, 1/EA
Supplier:  Foxx Life Sciences 3922004OEM
Encompass
392-2004-OEM: UN/DOT Container 20L HDPE with S70 Cap. 20L HDPE EZwaste UN/DOT System is designed to be an affordable solution for liquid waste disposal. Our containers are made to be tough and durable. They are great for secure shipment and storage of liquid products. The DOT/UN container is available in 13.5L, 20L and 60L. Each comes with a 70S cap. These containers can also be used in other industries including, but not limited to sanitation, chemical, agricultural and food. All raw materials used in our containers meet FDA Standards for food contact. Made from HDPE, meets UN 3H1 Standards.
Catalog No. 501050016
Only null left