missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Fumarate, USP, 97-101%, Spectrum™ Chemical

Catalog No. 18602698
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602698 50 kg Amber Glass Bottle
18602695 500 g Amber Glass Bottle
18602696 2.5 kg Amber Glass Bottle
18602697 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602698 Supplier Spectrum Chemical Mfg Cor Supplier No. FE10850KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

FE108, 141-01-5, C4H2FeO4

Spectrum™ Chemical Ferrous Fumarate, USP is used primarily to treat iron deficiency anemia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 141-01-5
Molecular Formula C4H2FeO4
Molecular Weight (g/mol) 169.90
InChI Key PMVSDNDAUGGCCE-ODZAUARKSA-L
IUPAC Name λ2-iron(2+) (2Z)-but-2-enedioate
SMILES [Fe++].[O-]C(=O)\C=C/C([O-])=O

Specifications

Physical Form Powder, Crystals and/or Chunks
CAS Max % 100%
Quantity 50 kg
Loss on Drying 1.5%
Packaging Amber Glass Bottle
Percent Purity 97 to 101%
Grade USP
Chemical Name or Material Ferrous Fumarate