missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ CBL, 16FT Serial DB9 TO Serial DIN8

Catalog No. 4316489
Encompass
Change view
Click to view available options
Quantity:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4316489 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4316489 Supplier Applied Biosystems™ Supplier No. 4316489

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Each package contains 16 ft of Serial DB9 to Serial DIN9 cable for the 310 Genetic Analyzer.

Each package contains 16 ft of Serial DB9 to Serial DIN9 cable for the 310 Genetic Analyzer.

Specifications

Quantity 1 unit
For Use With (Equipment) 310 Genetic Analyzer
Product Type Cable
Shipping Condition Room Temperature
Content And Storage Store at room temperature.

For Research Use Only. Not for use in diagnostic procedures.