Learn More
Applied Biosystems™ Cathode Buffer Container (CBC), for 3500/SeqStudio™ Flex

Description
The cathode buffer container (CBC) contains 1X running buffer to support all electrophoresis applications on Applied Biosystems 3500 and SeqStudio Flex series genetic analyzers. The container has two separate compartments, the left side provides the cathode buffer for electrophoresis and the right side provides for a capillary wash and spent polymer waste ejection functionality between injections.
The CBC is made in a ready-to-use, disposable container with a radio frequency identification (RFID) tag incorporated into the label. The top of the CBC is heat sealed with a plastic film, which should be removed prior to direct installation on to the instrument. Each package includes four individual containers.
Order Info
4 Pack
Specifications
Specifications
| Content And Storage | Contains 4 packs of cathode buffer container. Store the Cathode Buffer Container at 2-8°C until ready to use. Refer to the expiration date on the label. |
| For Use With (Equipment) | 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers |
| Type | Cathode Buffer Container |
| Quantity | 4 pack |
Frequently Asked Questions (FAQs)
Yes, it is still usable. We ship these buffers at room temperature to minimize the environmental impact of packaging and shipping these products on gel packs. We performed stability and performance studies and found that there was no impact to the performance of these buffers when shipped at room temperature. Please see the CE Buffers Ambient Shipping white paper detailing these studies, for your reference.
Some of the other causes of an Unstable Electrophoresis current detected error message are:
1.Leak on the system
2. Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3. Running Buffer that was:
-On the instrument longer than 2 weeks
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues
Inspect the system for leaks. If you do not see any leaks on the system, perform the wash the pump chamber and channels wizard using the conditioning pouch and place fresh, non-expired polymer and fresh Anode and Cathode buffer containers. If the problem persists, a service call may be required.
Putting the polymer back at 2-8 degrees C may extend the life of the polymer, however, we do not have any data indicating how long it might extend it. Removing the polymer and putting it back on the system can also result in a trade-off in usage. When placing it back on the system, you will need to degas the polymer for 30-60 min, refill the pump/block, array, and remove bubbles, which will cause you to use more polymer for maintenance than you would for running samples and may decrease the number of runs you can get out of a single polymer bottle/pouch.
If the instrument is not in use, it is not necessary to replace the buffer every 14 days. However, the buffer level needs to remain at the fill line to prevent the capillary or array from drying out. Top off the buffer volume with water if necessary. Replace the anode and cathode buffer containers prior to starting a new run.
For the Applied Biosystems 3500 Series Genetic Analyzer, the polymer can remain on the instrument for 14 days if room temperature is <25 degrees C and for 7 days if >25 degrees C.
For Research Use Only. Not for use in diagnostic procedures.