Learn More
Thermo Scientific Chemicals Cathepsin D, Human liver
Description
Cathepsin D, human liver is useful for inhibitor screening, kinetic studies, and cleavage of substrates in vitro or in tissue culture. It contains the mannose-6-phosphate lysosomal localization signal that targets the enzyme to the lysosomal compartment where it functions in the normal degradation of proteins. It is a useful prognostic indicator for breast cancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Species | Human |
| Purification Method | HPLC |
| Content And Storage | -30°C to -10°C |
| Format | Solid |
| Product Type | Cathepsin D, human liver |
| Quantity | 25 μg |
| Regulatory Status | RUO - research use only |
| Sequence | EC 3.4.23.5 |
RUO – Research Use Only