missing translation for 'onlineSavingsMsg'
Learn More

(+)-Catechin Hydrate 97.0+%, TCI America™

Catalog No. p-7124571
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07051G 1 g
C070510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07051G Supplier TCI America Supplier No. C07051G

Chemical Identifiers

CAS 154-23-4
Molecular Formula C15H14O6
Molecular Weight (g/mol) 290.271
MDL Number MFCD00075649
InChI Key PFTAWBLQPZVEMU-DZGCQCFKSA-N
Synonym (+)-3,3′C,4′C,5,7-Flavanpentol
PubChem CID 9064
ChEBI CHEBI:15600
IUPAC Name (2R,3S)-2-(3,4-dihydroxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol
SMILES C1C(C(OC2=CC(=CC(=C21)O)O)C3=CC(=C(C=C3)O)O)O

Specifications

Melting Point 96°C
Quantity 1 g
Formula Weight 290.27
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (+)-Catechin Hydrate
RTECSNumber DJ3450000
TSCA No