Learn More
Biomatik Corporation Recombinant Complement Component 5a (C5a), 100UG
Catalog No. 501505518
Encompass
Recombinant Rat Complement Component 5a (C5a) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Complement Component 5a (C5a). Accession Number: P08650. Expression Region: Gln12~Ile64. Tag Info: N-terminal His and GST Tag. Theoretical MW: 36kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 501505518 Supplier Biomatik Corporation Supplier No. RPU40952
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant Rat Complement Component 5a (C5a) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Complement Component 5a (C5a). Accession Number: P08650. Expression Region: Gln12~Ile64. Tag Info: N-terminal His and GST Tag. Theoretical MW: 36kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.