Catalase, Filtered, Minimum Activity: 40000U/mg Protein, Supplied as an aqueous solution of 2X crystallized catalase filtered through a 0.22u pore size Membrane, Store at 2 deg. to 8 deg.C, 10mL
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Catalase, Filtered, Minimum Activity: 40000U/mg Protein, Supplied as an aqueous solution of 2X crystallized catalase filtered through a 0.22u pore size Membrane, Store at 2 deg. to 8 deg.C, 10mL