missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Enespro™ CAT 2 Arc Flash Accessory Kit with Class 0 Gloves

Catalog No. 170011213
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
170011213 2X-Large
170011209 Small
170011210 Medium
170011211 Large
170011212 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 170011213 Supplier National Safety Apparel™ Supplier No. KIT2NC12PVB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

With a unique blend of 88% cotton and 12% high-tenacity nylon UltraSoft garments provide a smooth feel for increased comfort and the inherently FR material delivers exceptional performance in reliability

  • Flat locked seams for comfort
  • Rib knit, single layer hood balaclava
  • Adjustable settings allow the faceshield to be secured in various positions
  • Lightweight design
  • PureView™ technology for proven color recognition

Specifications

Certifications/Compliance CAT 2, NFPA 70E, CSA Z462, ASTM F1506, ASTM F2178, ANSI/ASTM D120, Class 0, Type 1, OSHA 1910.269
Material Westex™ UltraSoft™
Gender Unisex
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Product Type Accessory Kit
Arc Flash Rating 12 cal/cm²
Size 2X-Large
Suspender None
Color Navy
Closure Type None
Seam Type Stitched
Show More Show Less