Learn More
CritiCore CAT 1 Women's Lab Coat - Nomex™ FR
Catalog No. 17012877
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17012877 Small
17012878 Medium
17012879 Large
17012880 X-Large
17012881 2X-Large
17012882 3X-Large
17012883 4X-Large
17012884 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17012877 Supplier CritiCore Supplier No. 12L34950CXP63PSBLUE
Only null left
Lab Coat designed to provide protection against flash fire, electric arc (HRC 1), and other dangerous conditions. This Flame-Resistant lab coat is NFPA 2112:2023 Certified with numerous applications in both laboratory and production settings.
  • Fabric: Nomex™ FR
  • Blend: 100% Nomex™ IIIA,: prevents clothing ignition during short-duration thermal incidents and emergency exposure to flame
  • Arc Rating (ATPV) - 6.5 cal/sq cm
  • Quick-release snap front
  • Safety sleeve cuff with tunneled elastic
  • FR identification patches on sleeves and back
  • Excellent heat-resistance properties
  • Includes a waistband in the back for a more tailored fit
  • NFPA 2112:2023 Certified
Specifications
Product Type Flame-resistant Lab Coat
Size Small
Gender Women's
Material Nomex
Color Blue
Closure Type Snaps
Cuff Style Elastic
Disposable Reusable
Sterility Non-sterile
Resistance Flame
Style Standard Length
No. of Pockets 3
Construction Sewn
Show More Show Less