missing translation for 'onlineSavingsMsg'
Learn More

Castor oil

CAS: 8001-79-4 | C57H104O9 | 933.45 g/mol

Supplier:  Thermo Scientific Chemicals 404160025

Quantity 2.5 kg

 View more versions of this product

Catalog No. AC404160025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

8001-79-4
MFCD00130879

Specifications

Castor oil
-10°C
0.9570g/mL
229°C
Glass bottle
2.5 kg
176 to 182g KOH/g
0.957
Solubility in water: .. Other solubilities: miscible with alcohol,benzene,chloroform,,carbondisulfide,ether and glacial acetic acid
Viscous Liquid
8001-79-4
Yellow
229°C
Authentic
1.4770 to 1.481
MFCD00130879
14,1898
+ 3.5
0,6-0,8 Pas (25°C)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-293-8

RTECSNumber : FI4100000

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions