missing translation for 'onlineSavingsMsg'
Learn More

Castor oil

Catalog No. AC397282500
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC397282500 250 g
AC404165000 500 g
AC397280010 1 kg
AC404160025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC397282500 Supplier Thermo Scientific Chemicals Supplier No. 397282500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 8001-79-4 | C57H104O9 | 933.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 8001-79-4
Molecular Formula C57H104O9
Molecular Weight (g/mol) 933.45
MDL Number MFCD00130879
InChI Key ZEMPKEQAKRGZGQ-UHFFFAOYNA-N
IUPAC Name 1,3-bis[(12-hydroxyoctadec-9-enoyl)oxy]propan-2-yl 12-hydroxyoctadec-9-enoate
SMILES CCCCCCC(O)CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(O)CCCCCC)OC(=O)CCCCCCCC=CCC(O)CCCCCC

Specifications

Melting Point 4.0°C
Color Yellow
Density 1.0500g/mL
Boiling Point >100.0°C
Flash Point 257°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 250 g
Saponification Value 176 to 182g KOH/g
Specific Gravity 1.05
Solubility Information Solubility in water: 100g/L (20°C). Other solubilities: soluble in organic solvents
Viscosity 650-800 mPa.s (25°C)
Physical Form Viscous Liquid
Chemical Name or Material Castor oil
Show More Show Less
EINECSNumber 500-151-7
RTECSNumber FI4100000
TSCA TSCA

RUO – Research Use Only