Learn More
Epredia™ Cassette II Slotted Tissue Cassettes in Tube Packs
Slotted Cassettes are ideal for routine tissue samples. They are compatible with all Epredia PrintMate™ Cassette Printers, have a front edge lid hinge, and loose-packed lids that are available in 12 color varieties.
Supplier:  Epredia™ B851729BL
 View more versions of this product
Catalog No. B851729BL
Only null left
Description
Description
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
Specifications
Chemical Identifiers
Yes
Slots
Yes
Cassette II Biopsy Cassette
750/Cs.
Separate Lid, Front Hinge
For Use With PrintMate™ Printers
Specifications
Slots
Yes
Yes
For Use With PrintMate™ Printers
750/Cs.
Separate Lid, Front Hinge
Blue
Cassette II Biopsy Cassette
Product Suggestions
Product Suggestions