missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Cassette II Slotted Tissue Cassettes in Tube Packs

Catalog No. B851729BE
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
B851729BE Beige
B851729BL Blue
B851729GN Green
B851729GY Gray
B851729LA Lavender
B851729LG Light Green
B851729LI Lilac
B851729OR Orange
B851729TN Tan
B851729WH White
B851729YW Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. B851729BE Supplier Epredia™ Supplier No. B851729BE

Slotted Cassettes are ideal for routine tissue samples. They are compatible with all Epredia PrintMate™ Cassette Printers, have a front edge lid hinge, and loose-packed lids that are available in 12 color varieties.

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime

Grossing, Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving

Specifications

Pore Style Slots
Quantity 750/Cs.
Closure Included Yes
Closure Type Separate Lid, Front Hinge
PrintMate Compatible Yes
Color Beige
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type Cassette II Biopsy Cassette