Learn More
Epredia™ Cassette II Biopsy Tissue Cassettes
For small tissue samples, these cassettes have a front-edge lid hinge with loose-packed lids that are available in 12 different color varieties
Supplier:  Epredia™ B851738LA
 View more versions of this product
Catalog No. B851738LA
Only null left
Explore available promotions
Description
Description
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
Specifications
Specifications
Small Squares
1000/Cs.
Separate Lid, Front Hinge
Lavender
Yes
Loose Packed (Lids)
No
Cassette II with Color-Matched Lids
Product Suggestions
Product Suggestions