missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Cassette Foam Sponges

Catalog No. 45001956
Change view
Click to view available options
Thickness (English):
0.11 in.
Thickness (Metric):
3 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thickness (English) Thickness (Metric)
45001956 0.11 in. 3 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45001956 Supplier Cytiva Supplier No. WHTTE25F1/8
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

These Cassette Foam Sponges are accessory items for transfer tanks.

  • Cassette foam sponges

Specifications

For Use With (Equipment) TE 22 Mini Tank Transfer Unit
Type Cassette Foam Sponge
Material Foam
Quantity 4 Pk.
Thickness (English) 0.11 in.
Thickness (Metric) 3 mm