Learn More
Epredia™ Cassette 1 Slotted Tissue Cassettes
Catalog No. 1000962
Change view
Click to view available options
Color:
Blue
Green
Peach
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
1000962 Peach
1000957 White
1000959 Green
1000960 Yellow
1000961 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 1000962 Supplier Epredia™ Supplier No. 1000962
Hold tissue specimens during processing and embedding with Epredia™ Cassette 1 slotted tissue cassettes.
  • Acetal polymer construction
  • 30° writing face
  • Small slot-style pores
Ordering Alerts:
  • Supplied in box of 500 cassettes with attached breakaway lids (not available in Tube Packs)
Specifications
Pore Style Slotted Pore
Writing Face Angle 30°
Color Peach
Material Acetal Polymer
Product Type Tissue Cassette