missing translation for 'onlineSavingsMsg'
Learn More

Promega Caspase-Glo™ 3/7 Assay Kit

Catalog No. p-2626200
Encompass
Change view
Click to view available options
Quantity:
2.5 mL
10 mL
100 mL
10 x 10 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG8091 10 mL
PRG8090 2.5 mL
PRG8092 100 mL
PRG8093 10 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PRG8091 Supplier Promega Supplier No. G8091

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Caspase-Glo 3/7 Substrate; Caspase-Glo 3/7 Buffer

Caspase-Glo™ 3/7 Assay is a homogeneous luminescent assay that measures caspase-3/7 activities

  • Apoptosis Detection with Unmatched Speed and Sensitivity
  • High sensitivity—requires fewer cells and less enzyme
  • Simple add-mix-measure protocol—no sample preparation
  • Easily scalable to 96-, 384- and 1,536-well plate formats

  • Patent pending.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patent Nos. 5,583,024; 5,674,713; and 5,700,673.

Specifications

Quantity 10 mL
pH 7.4
Test Time 0.25 to 1 hr.
Content And Storage 1 x 10mL Caspase-Glo™ 3/7 Buffer, 1 x 1 bottle Caspase-Glo™ 3/7 substrate. Store at -30°C to -10°C
For Use With (Application) 96- or 384-well plates using either purified enzyme or cells in culture as sample
Form Liquid