missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propylene Glycol Methyl Ether, 97%, Spectrum™ Chemical
Specifications
Specifications
| Boiling Point | 120°C (248°F) |
| CAS Max % | 1 |
| Quantity | 4 L |
| Packaging | Amber Glass Bottle |
| Water | 0.001 |
| Molecular Weight (g/mol) | 90.12 |
| Percent Purity | 97% |
| Grade | Ungraded |
| Chemical Name or Material | Propylene Glycol Methyl Ether |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.