missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Caspase-1 Inhibitor IV

Catalog No. AAJ64583MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64583MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64583MCR Supplier Thermo Scientific Chemicals Supplier No. J64583MCR

 

A competitive, cell-permeable, and irreversible inhibitor of caspases -1, -4, and -5

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 154674-81-4
Molecular Formula C33H42N4O10
Molecular Weight (g/mol) 654.72
MDL Number MFCD00798794
InChI Key LOACDHZQOPEFKK-UHFFFAOYNA-N
IUPAC Name 5-(2,6-dimethylbenzoyloxy)-3-(2-{2-[2-acetamido-3-(4-hydroxyphenyl)propanamido]-3-methylbutanamido}propanamido)-4-oxopentanoic acid
SMILES CC(C)C(NC(=O)C(CC1=CC=C(O)C=C1)NC(C)=O)C(=O)NC(C)C(=O)NC(CC(O)=O)C(=O)COC(=O)C1=C(C)C=CC=C1C

Specifications

Content And Storage Store at -20°C
Format Powder
Quantity 1 mg

RUO – Research Use Only