missing translation for 'onlineSavingsMsg'
Learn More

Formamide, Deionized, Ultrapure, 99.5%, Spectrum™ Chemical

Catalog No. 18602837
Change view
Click to view available options
Quantity:
500 mL
950 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602837 500 mL -
18602836 950 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18602837 Supplier Spectrum Chemical Mfg Cor Supplier No. F3425500ML
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

F3425, 75-12-7, CH3NO

Spectrum™ Chemical Formamide, Deionized, Ultrapure - Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates. It is used as a RNA stabilizer in gel electrophoresis by deionizing RNA, in capillary electrophoresis to stabilize (single) strands of denatured DNA and as a constituent of cryoprotectant vitrification mixtures used for cryopreservation of tissues and organs.

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
IUPAC Name formamide
SMILES NC=O

Specifications

CAS Max % 100%
Boiling Point 211°C/411.8°F
Quantity 500 mL
Percent Purity 99.5%
Grade Ultra Pure
Physical Form Liquid
Chemical Name or Material Formamide, Deionized