missing translation for 'onlineSavingsMsg'
Learn More

Casein, ('Purified), MP Biomedicals

Catalog No. ICN90129301
Change view
Click to view available options
Quantity:
1 lb.
25 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN90129301 1 lb.
ICN90129325 25 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN90129301 Supplier MP Biomedicals Inc Supplier No. 0290129301

Chemical Identifiers

CAS 9000-71-9
Molecular Formula C81H125N22O39P
Molecular Weight (g/mol) 2061.98
MDL Number MFCD00081481
InChI Key BECPQYXYKAMYBN-UHFFFAOYNA-N
Synonym casein, caseinogen, casein, tech., bovine beta-casein monophosphopeptide
PubChem CID 73995022
IUPAC Name 6-amino-2-{[2-({2-[(2-{[2-({2-[(2-{[2-({2-[(2-{[2-({2-[(2-{[2-({2-[(2-amino-1-hydroxy-3-phenylpropylidene)amino]-1-hydroxy-4-(C-hydroxycarbonimidoyl)butylidene}amino)-1-hydroxy-3-(phosphonooxy)propylidene]amino}-4-carboxy-1-hydroxybutylidene)amino]-4-carboxy-1-hydroxybutylidene}amino)-1-hydroxy-4-(C-hydroxycarbonimidoyl)butylidene]amino}-1-hydroxy-4-(C-hydroxycarbonimidoyl)butylidene)amino]-1-hydroxy-4-(C-hydroxycarbonimidoyl)butylidene}amino)-1,3-dihydroxybutylidene]amino}-4-carboxy-1-hydroxybutylidene)amino]-3-carboxy-1-hydroxypropylidene}amino)-4-carboxy-1-hydroxybutylidene]amino}-1-hydroxy-4-methylpentylidene)amino]-1-hydroxy-4-(C-hydroxycarbonimidoyl)butylidene}amino)-3-carboxy-1-hydroxypropylidene]amino}hexanoic acid
SMILES CC(C)CC(N=C(O)C(CCC(O)=O)N=C(O)C(CC(O)=O)N=C(O)C(CCC(O)=O)N=C(O)C(N=C(O)C(CCC(O)=N)N=C(O)C(CCC(O)=N)N=C(O)C(CCC(O)=N)N=C(O)C(CCC(O)=O)N=C(O)C(CCC(O)=O)N=C(O)C(COP(O)(O)=O)N=C(O)C(CCC(O)=N)N=C(O)C(N)CC1=CC=CC=C1)C(C)O)C(O)=NC(CCC(O)=N)C(O)=NC(CC(O)=O)C(O)=NC(CCCCN)C(O)=O

Specifications

Quantity 1 lb.
Grade Purified
Chemical Name or Material Casein, High Nitrogen
Recommended Storage Room Temperature