Learn More
New England Biolab Ltd Cas9 Nuclease, S. pyogenes 400 pmol
Catalog No. 501526391
Encompass
Cas9 Nuclease, S. pyogenes, is an RNA-guided endonuclease that catalyzes site-specific cleavage of double stranded DNA. The location of the break is within the target sequence 3 bases from the NGG PAM (Protospacer Adjacent Motif) . The PAM sequence, NGG, must follow the targeted region on the opposite strand of the DNA with respect to the region complementary sgRNA sequence.
Catalog No. 501526391 Supplier New England Biolab Ltd Supplier No. M0386T
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Cas9 Nuclease, S. pyogenes, is an RNA-guided endonuclease that catalyzes site-specific cleavage of double stranded DNA. The location of the break is within the target sequence 3 bases from the NGG PAM (Protospacer Adjacent Motif) . The PAM sequence, NGG, must follow the targeted region on the opposite strand of the DNA with respect to the region complementary sgRNA sequence.