missing translation for 'onlineSavingsMsg'
Learn More

(-)-Carveol, mixture of isomers, 97%

CAS: 99-48-9 | C10H16O | 152.24 g/mol

Supplier:  Thermo Scientific Chemicals B2063618

 View more versions of this product

Catalog No. AAB2063618

Only null left
Explore available promotions

Description

Description

It is a terpene that inhibits mammary carcinogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a terpene that inhibits mammary carcinogenesis.

Solubility
Partly soluble in water. Soluble in alcohol, ethanol and DMSO.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications

Specifications

(-)-Carveol
99-48-9
227°C
C10H16O
MFCD00869995,MFCD00062993
6269416
Partly soluble in water. Soluble in alcohol,ethanol and DMSO.
CC(=C)C1CC=C(C)C(O)C1
152.24
CHEBI:23046
97%
mixture of isomers
0.96
98°C (209°F)
1.496
50 g
carveol, p-mentha-6,8-dien-2-ol, p-mentha-1,8-dien-6-ol, 1-methyl-4-isopropenyl-6-cyclohexen-2-ol, 2-cyclohexen-1-ol, 2-methyl-5-1-methylethenyl, p-mentha-1 6 ,8-dien-2-ol, 5-isopropenyl-2-methyl-2-cyclohexen-1-ol, --carveol, l-carveol, 2-methyl-5-1-methylethenyl-2-cyclohexen-1-ol
BAVONGHXFVOKBV-UHFFFAOYNA-N
2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-ol
7438
152.24

Chemical Identifiers

99-48-9
152.24
BAVONGHXFVOKBV-UHFFFAOYNA-N
7438
2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-ol
C10H16O
MFCD00869995,MFCD00062993
carveol, p-mentha-6,8-dien-2-ol, p-mentha-1,8-dien-6-ol, 1-methyl-4-isopropenyl-6-cyclohexen-2-ol, 2-cyclohexen-1-ol, 2-methyl-5-1-methylethenyl, p-mentha-1 6 ,8-dien-2-ol, 5-isopropenyl-2-methyl-2-cyclohexen-1-ol, --carveol, l-carveol, 2-methyl-5-1-methylethenyl-2-cyclohexen-1-ol
CHEBI:23046
CC(=C)C1CC=C(C)C(O)C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H320-H335

EINECSNumber : 202-757-4

RTECSNumber : OS8400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only