missing translation for 'onlineSavingsMsg'
Learn More

(-)-Carveol, mixture of isomers, 97%

Catalog No. AAB2063609
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2063609 10 g
AAB2063618 50 g
AAB2063630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2063609 Supplier Thermo Scientific Chemicals Supplier No. B2063609
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 99-48-9 | C10H16O | 152.24 g/mol

It is a terpene that inhibits mammary carcinogenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a terpene that inhibits mammary carcinogenesis.

Solubility
Partly soluble in water. Soluble in alcohol, ethanol and DMSO.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 99-48-9
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00869995,MFCD00062993
InChI Key BAVONGHXFVOKBV-UHFFFAOYNA-N
Synonym carveol, p-mentha-6,8-dien-2-ol, p-mentha-1,8-dien-6-ol, 1-methyl-4-isopropenyl-6-cyclohexen-2-ol, 2-cyclohexen-1-ol, 2-methyl-5-1-methylethenyl, p-mentha-1 6 ,8-dien-2-ol, 5-isopropenyl-2-methyl-2-cyclohexen-1-ol, --carveol, l-carveol, 2-methyl-5-1-methylethenyl-2-cyclohexen-1-ol
PubChem CID 7438
ChEBI CHEBI:23046
IUPAC Name 2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-ol
SMILES CC(=C)C1CC=C(C)C(O)C1

Specifications

Density 0.96
Boiling Point 227°C
Flash Point 98°C (209°F)
Refractive Index 1.496
Quantity 10 g
Beilstein 6269416
Solubility Information Partly soluble in water. Soluble in alcohol,ethanol and DMSO.
Formula Weight 152.24
Percent Purity 97%
Chemical Name or Material (-)-Carveol, mixture of isomers
Hazard Category H315-H320-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-757-4
RTECSNumber OS8400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only