missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carvedilol 98.0+%, TCI America™
$329.94 - $1178.08
Chemical Identifiers
| CAS | 72956-09-3 |
|---|---|
| Molecular Formula | C24H26N2O4 |
| Molecular Weight (g/mol) | 406.48 |
| MDL Number | MFCD00864692,MFCD00869663 |
| InChI Key | OGHNVEJMJSYVRP-UHFFFAOYNA-N |
| Synonym | carvedilol, coreg, dilatrend, carvedilolum, eucardic, carvedilolum latin, kredex, querto, 1-9h-carbazol-4-yloxy-3-2-2-methoxyphenoxy ethyl amino-2-propanol, 1-9h-carbazol-4-yl oxy-3-2-2-methoxyphenoxy ethyl amino propan-2-ol |
| PubChem CID | 2585 |
| ChEBI | CHEBI:3441 |
| IUPAC Name | 1-(9H-carbazol-4-yloxy)-3-{[2-(2-methoxyphenoxy)ethyl]amino}propan-2-ol |
| SMILES | COC1=CC=CC=C1OCCNCC(O)COC1=CC=CC2=C1C1=C(N2)C=CC=C1 |
Chemical Identifiers
| 72956-09-3 | |
| 406.48 | |
| OGHNVEJMJSYVRP-UHFFFAOYNA-N | |
| 2585 | |
| 1-(9H-carbazol-4-yloxy)-3-{[2-(2-methoxyphenoxy)ethyl]amino}propan-2-ol |
| C24H26N2O4 | |
| MFCD00864692,MFCD00869663 | |
| carvedilol, coreg, dilatrend, carvedilolum, eucardic, carvedilolum latin, kredex, querto, 1-9h-carbazol-4-yloxy-3-2-2-methoxyphenoxy ethyl amino-2-propanol, 1-9h-carbazol-4-yl oxy-3-2-2-methoxyphenoxy ethyl amino propan-2-ol | |
| CHEBI:3441 | |
| COC1=CC=CC=C1OCCNCC(O)COC1=CC=CC2=C1C1=C(N2)C=CC=C1 |
Specifications
| 72956-09-3 | |
| White-Yellow | |
| MFCD00864692,MFCD00869663 | |
| 3077 | |
| OGHNVEJMJSYVRP-UHFFFAOYNA-N | |
| 1-(9H-carbazol-4-yloxy)-3-{[2-(2-methoxyphenoxy)ethyl]amino}propan-2-ol | |
| 2585 | |
| 406.48 | |
| Crystalline Powder |
| 117°C | |
| C24H26N2O4 | |
| 5 g | |
| carvedilol, coreg, dilatrend, carvedilolum, eucardic, carvedilolum latin, kredex, querto, 1-9h-carbazol-4-yloxy-3-2-2-methoxyphenoxy ethyl amino-2-propanol, 1-9h-carbazol-4-yl oxy-3-2-2-methoxyphenoxy ethyl amino propan-2-ol | |
| COC1=CC=CC=C1OCCNCC(O)COC1=CC=CC2=C1C1=C(N2)C=CC=C1 | |
| 406.48 | |
| CHEBI:3441 | |
| ≥98.0% (HPLC,T) | |
| Carvedilol |
Safety and Handling
RTECSNumber : UA8670000
TSCA : No