missing translation for 'onlineSavingsMsg'
Learn More

Carvedilol 98.0+%, TCI America™

Catalog No. C22605G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22605G 5 g
C226025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22605G Supplier TCI America Supplier No. C22605G
Only null left

Chemical Identifiers

CAS 72956-09-3
Molecular Formula C24H26N2O4
Molecular Weight (g/mol) 406.48
MDL Number MFCD00864692,MFCD00869663
InChI Key OGHNVEJMJSYVRP-UHFFFAOYNA-N
Synonym carvedilol, coreg, dilatrend, carvedilolum, eucardic, carvedilolum latin, kredex, querto, 1-9h-carbazol-4-yloxy-3-2-2-methoxyphenoxy ethyl amino-2-propanol, 1-9h-carbazol-4-yl oxy-3-2-2-methoxyphenoxy ethyl amino propan-2-ol
PubChem CID 2585
ChEBI CHEBI:3441
IUPAC Name 1-(9H-carbazol-4-yloxy)-3-{[2-(2-methoxyphenoxy)ethyl]amino}propan-2-ol
SMILES COC1=CC=CC=C1OCCNCC(O)COC1=CC=CC2=C1C1=C(N2)C=CC=C1

Specifications

Melting Point 117°C
Color White-Yellow
Quantity 5 g
UN Number 3077
Formula Weight 406.48
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Carvedilol
RTECSNumber UA8670000
TSCA No