missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carvacrol 98.0+%, TCI America™
$44.28 - $394.66
Chemical Identifiers
| CAS | 499-75-2 |
|---|---|
| Molecular Formula | C10H14O |
| Molecular Weight (g/mol) | 150.22 |
| MDL Number | MFCD00002236 |
| InChI Key | RECUKUPTGUEGMW-UHFFFAOYSA-N |
| Synonym | carvacrol, 5-isopropyl-2-methylphenol, isopropyl-o-cresol, o-thymol, antioxine, isothymol, karvakrol, 5-isopropyl-o-cresol, 2-p-cymenol, 2-hydroxy-p-cymene |
| PubChem CID | 10364 |
| ChEBI | CHEBI:3440 |
| IUPAC Name | 2-methyl-5-(propan-2-yl)phenol |
| SMILES | CC(C)C1=CC=C(C)C(O)=C1 |
Chemical Identifiers
| 499-75-2 | |
| 150.22 | |
| RECUKUPTGUEGMW-UHFFFAOYSA-N | |
| 10364 | |
| 2-methyl-5-(propan-2-yl)phenol |
| C10H14O | |
| MFCD00002236 | |
| carvacrol, 5-isopropyl-2-methylphenol, isopropyl-o-cresol, o-thymol, antioxine, isothymol, karvakrol, 5-isopropyl-o-cresol, 2-p-cymenol, 2-hydroxy-p-cymene | |
| CHEBI:3440 | |
| CC(C)C1=CC=C(C)C(O)=C1 |
Specifications
| 499-75-2 | |
| 238°C | |
| MFCD00002236 | |
| carvacrol, 5-isopropyl-2-methylphenol, isopropyl-o-cresol, o-thymol, antioxine, isothymol, karvakrol, 5-isopropyl-o-cresol, 2-p-cymenol, 2-hydroxy-p-cymene | |
| CC(C)C1=CC=C(C)C(O)=C1 | |
| 150.22 | |
| CHEBI:3440 | |
| ≥98.0% (GC) | |
| Carvacrol |
| Yellow | |
| C10H14O | |
| 25 g | |
| RECUKUPTGUEGMW-UHFFFAOYSA-N | |
| 2-methyl-5-(propan-2-yl)phenol | |
| 10364 | |
| 150.22 | |
| Liquid |
Safety and Handling
EINECSNumber : (4)-0057
RTECSNumber : FI1225000
TSCA : Yes