missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Carteolol hydrochloride

Catalog No. AC464320050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464320050 5 g
AC464320010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464320050 Supplier Thermo Scientific Chemicals Supplier No. 464320050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51781-21-6
Molecular Formula C16H25ClN2O3
Molecular Weight (g/mol) 328.84
MDL Number MFCD00941499
InChI Key FYBXRCFPOTXTJF-UHFFFAOYNA-N
IUPAC Name hydrogen 5-[3-(tert-butylamino)-2-hydroxypropoxy]-1,2,3,4-tetrahydroquinolin-2-one chloride
SMILES [H+].[Cl-].CC(C)(C)NCC(O)COC1=CC=CC2=C1CCC(=O)N2

Specifications

Melting Point 278.0°C
Quantity 5 g
Solubility Information Solubility aq. soln.: soluble. Other solubilities: moderately soluble in methanol. slightly soluble, in acetic acid. insoluble in chloroform, ether,, dichloromethane
Formula Weight 328.83
Chemical Name or Material Carteolol hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes serious eye irritation.

Suspected of damaging the unborn child.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

If eye irritation persists: Get medical advice/attention.

Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber 257-415-7

RUO – Research Use Only