missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Carson's Modified Buffered Formalin

Catalog No. 57141
Change view
Click to view available options
Capacity:
5 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
57141 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 57141 Supplier Epredia™ Supplier No. 57141

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Performs similarly to 10% neutral buffered formalin and is an excellent general-use fixative. 5-gallon size.

  • Epredia Carson's Modified Buffered Formalin is an excellent general-use fixative
  • Performs similarly to 10% neutral buffered formalin
  • Phosphate buffers are used to stabilize the pH between 6.8 and 7.2
  • Use for electron microscopy specimens

Specifications

Capacity 5 gal.
For Use With (Equipment) All Tissue Processors
Grade Histological
Product Type Formalin
Solution Type Formalin
pH 6.8 to 7.2
Form Liquid

Non-sterile