missing translation for 'onlineSavingsMsg'
Learn More

CleanSpace™ Technology Carry Backpack

Catalog No. 17800626
Change view
Click to view available options
For Use With (Equipment):
Respirators
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
17800626 Respirators
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17800626 Supplier CleanSpace™ Technology Supplier No. PAF0099

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Carry backpack for use with CleanSpace™ respirator systems.

  • Black
  • For use with all CleanSpace™ Respirators

Specifications

Color Black
Disposable Reusable
Product Type Backpack
For Use With (Equipment) Respirators
Model PAF-0099