missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Carrier RNA

Catalog No. 4382878
Encompass
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4382878 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4382878 Supplier Applied Biosystems™ Supplier No. 4382878

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ambion™ Carrier RNA (1 mg/mL) is a component of all the MagMAX™ Isolation Kits that may be purchased separately.

Provided in one tube of 500 μL.

Specifications

Content And Storage Store solution at –20°C.
For Use With (Application) Carrier for Nucleic Acid Precipitation
Product Line Ambion
Product Type Carrier RNA
Quantity 500 μL
Shipping Condition Dry Ice
I ran out of carrier RNA for my MagMAX isolation kit. Can I order this part separately?

Yes, carrier RNA can be ordered separately. The Cat. No. is 4382878.

For Research Use Only. Not for use in diagnostic procedures.