missing translation for 'onlineSavingsMsg'
Learn More

Caronic Anhydride 98.0+%, TCI America™

Catalog No. p-7125548
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25961G 1 g
C25965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25961G Supplier TCI America Supplier No. C25961G
Only null left

Chemical Identifiers

CAS 67911-21-1
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD09751198
InChI Key QKAHKEDLPBJLFD-UHFFFAOYSA-N
PubChem CID 10374536
IUPAC Name 6,6-dimethyl-3-oxabicyclo[3.1.0]hexane-2,4-dione
SMILES CC1(C2C1C(=O)OC2=O)C

Specifications

Melting Point 55°C
Color White
Boiling Point 137°C
Quantity 1 g
Synonym 6,6-dimethyl-3-oxabicyclo 3.1.0 hexane-2,4-dione, caronic anhydride, 3-oxabicyclo 3.1.0 hexane-2,4-dione, 6,6-dimethyl, 3,3-dimethyl-1,2-cyclopropanedicarboxylic anhydride, ksc626s3r, 6,6-dimethyl-3-oxabicyclo 3.1.0 hex-2,4-dione, 3,3-dimethyl-1,2-cyclopropane dicarboxylic anhydride, 3-oxabicyclo 3.1.0 hexane-2,4-dione,6,6-dimethyl, 3,3-dimethyl-1,2-cyclopropanedicarboxylic acid anhydride, 2r,4r-methyl 4-hydroxypyrrolidine-2-carboxylate hydrochloride form
Formula Weight 140.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Caronic Anhydride